AATB1408EA
442
EUR
InStock
AATB1408
iFluor® 860, Maleimide
iFluor®
In vivo fluorescence imaging uses a sensitive camera to detect fluorescence emission from fluorophores in whole-body living small animals.
- Bright and photostable
- Minimal quenching
- Stable conjugation