AATB2237EA
295
EUR
InStock
AATB2237
Tide Fluor™ 1 alkyne, Tide Fluor™ sonde fluorescente
Tide Fluor™ 1 alkyne
TF1 is designed to be a superior fluorophore alternative to EDANS.
- Stronger absorption and fluorescence intensity
- Lower environment-sensitive fluorescence
- pH-independent fluorescence from 3-11
- Improved sensitivity