AATB2274EA
590
EUR
InStock
AATB2274
Tide Fluor™ 3 phosphoramidite, Tide Fluor™ sonde fluorescente
Tide Fluor™ 3 phosphoramidite
Tide Fluor™ 3 (TF3) phosphoramidite is a high-performance alternative to Cy3 phosphoramidite for the in-synthesis labeling of oligonucleotides.
- Stronger absorption and fluorescence intensity
- Lower environment-sensitive fluorescence
- pH-independent fluorescence from 3-11
- Improved sensitivity