AATB2292EA
295
EUR
InStock
AATB2292
Tide Fluor™ 6WS amine, Tide Fluor™ sonde fluorescente
Tide Fluor™ 6WS amine
Tide Fluor™ 6WS (TF6WS) family has the spectral properties similar to those of Cy5.5, IRDye 700 and Alexa Fluor 680.
- Stronger absorption and fluorescence intensity
- Lower environment-sensitive fluorescence
- pH-independent fluorescence from 3-11
- Improved sensitivity