AATB2346EA
442
EUR
InStock
AATB2346
Tide Fluor™ 3WS succinimidyl ester, Tide Fluor™ sonde fluorescente
Tide Fluor™ 3WS succinimidyl ester
Tide Fluor™ 3WS (TF3WS) family has the spectral properties essentially identical to those of Cy3.
- Stronger absorption and fluorescence intensity
- Lower environment-sensitive fluorescence
- pH-independent fluorescence from 3-11
- Improved sensitivity