AATB4533EA
146
EUR
InStock
AATB4533
Propargyl-PEG5-NHS ester for biomolecular labeling
Propargyl-PEG5-NHS ester
Alkyne Modifier II, NHS ester is an alkyne-containing reagent that reacts with primary amines at pH 7 to 9 forming a stale amide bond.
- Improved crosslinking efficiency
- High water solubility