AATB80050EA
146
EUR
InStock
AATB80050
Staurosporine 1 mM, ReadiUse™ for apoptosis positive control
Staurosporine
Staurosporine is widely used as a positive control for inducing apoptosis.
- Well-cited positive apoptosis control
- Simple effective apoptosis induction in multiple cell lines
- Cell permeable and reversible
- Inhibits some types of platelet aggregation