Synonyms:
5-Bromopentanenitrile
L03952.06
L03952.14
L03952.06EA
55.2
EUR
InStock
L03952.06
L03952.14
5-Bromopentanenitrile ≥98%
5-Bromopentanenitrile
Purity: 98+%
CAS No.: 5414-21-1
Molecular Formula: Br(CH2)4CN
Formula Weight: 162.04
Flash Point: 110°(230°F)
Refractive Index: 1.4785
Boiling Point: 110-111°/12mm
Beilstein Reference: 1742080
UN No.: 3276
MDL No.: MFCD00001976