L11536.06
L11536.14
L11536.03
L11536.06EA
124
EUR
InStock
L11536.06
L11536.14
L11536.03
2-Mercaptobenzyl alcohol tech. 90%, grade Technique
2-Mercaptobenzyl alcohol
Purity: 90%
CAS No.: 4521-31-7
Molecular Formula: C7H8OS
Formula Weight: 140.21
Flash Point: 110°(230°F)
Refractive Index: 1.6230
Odour: Like rotten eggs
Melting Point: 31-32°
Boiling Point: 94-96°/0.2mm
Beilstein Reference: 2206253
MDL No.: MFCD00014448