Incubator Shakers, LT-X
Fournisseur: KUEHNER ADOLF
108671 108672
KUEH108671EA 22820 EUR InStock
KUEH108671 KUEH108672
Incubator Shakers, LT-X
Agitateurs Agitateurs incubateurs
The LT-X (Lab-Therm) incubator shaker has the same capabilities as the larger ISF1-Z and ISF4-X models. The incubator shaker was specially developed to address the shaking needs of the biotechnology and pharmaceutical industries. The practical size of the LT-X means it will fit in any laboratory.
  • Fits into any laboratory
  • Used in biotech and pharmaceutical industries
  • Accepts flasks up to 6 L
  • Two units can be stacked without the need for special kits or tools
  • Five years warranty
The LT-X can be equipped with options for CO₂, O₂, controlled humidity, LED photosynthesis, UV lighting and much more. It is now also possible to integrate measuring data from Kuhner shakers with your company’s own network using Kuhner Insight software together with the NET-60 interface.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité