WHEATON® Crimpenstein Crimpers and Decappers Jaw Set
Fournisseur: DWK Life Sciences
WHEATON® Crimpenstein
WHEAW225623EA 359 EUR InStock
WHEAW225642 WHEAW225623
WHEATON® Crimpenstein Crimpers and Decappers Jaw Set
Pinces à sertir et pinces à dessertir
The WHEATON Crimpenstein crimper and decapper jaw and jaw set are designed for efficiently sealing and opening 20 mm flip-cap aluminium seals in laboratory settings.
  • Crafted with precision and durability
  • Provides consistency in uniform crimps minimising seal breakge and leakage
  • Ergonomic design fo user comfort
  • Efficienent laboratory results
Ideal for use in pharmaceutical, chemical, or research laboratories, the WHEATON Crimpenstein crimper and decapper jaw set offers a dependable solution for sealing and opening containers with 20 mm flip-cap aluminium seals. Upgrade your sealing process with this reliable and user-friendly tool for enhanced productivity and reliable sample protection.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité