Endeavor™ 5000 Light Duty Digital Orbital Shakers
Fournisseur: Ohaus
Endeavor™
OHAU30770916EA 1010 EUR InStock
OHAU30770916
Endeavor™ 5000 Light Duty Digital Orbital Shakers
Agitateurs Agitateurs orbitaux
Digital orbital shakers you can trust for everyday shaking.
  • Budget-friendly: No need to buy a separate tray
  • Ready to attach a wide variety of accessories - customize for low to mid volume applications
  • Programmable: Save time and guarantee reproducible results
  • Simple on-board menu to set, store, and recall multi-step programs
  • Data logging and control: Remote function permits PC control and data transfer
Endeavor™ 5000 Light duty shakers is 20 mm orbit, or a dedicated microplate model. It can be used in incubators and cold rooms. Stable, accurate, long-term shaking is ensured with the triple eccentric drive and maintenance-free brushless DC motor. Integrated overload and unbalanced detection to protect samples.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité