Anti-Goat Bovine Polyclonal Antibody (CF®583R)
Fournisseur: Biotium
20888-500UL 20888-50UL
20888-500ULEA 248 EUR InStock
20888-500UL 20888-50UL
Anti-Goat Bovine Polyclonal Antibody (CF®583R)
Anticorps
Highly cross-adsorbed bovine anti-goat IgG (H L) secondary antibody labeled with our superior CF® Dyes.
  • Highly cross-adsorbed for specific staining with minimal background
  • Minimal cross-reactivity with milk or BSA blocking solutions
  • Available in 7 bright and photostable CF® Dyes
  • Suitable for western, immunofluorescence, and immunohistology
  • CF® Dye and biotin conjugates: add 0.5 ml distilled water
  • HRP or DNP conjugates: add 1 ml distilled water
Bovine serum albumin (BSA) and dry milk are used extensively for blocking non-specific binding of antibodies and for stabilizing antibodies and other proteins during freeze drying and in dilute solutions. However dry milk and most commercial sources of BSA contain bovine IgG which interferes with the use of anti-goat (IgG) secondary antibodies due to the close phylogenetic relationship between cows and goats.
Type: Secondary
Antigen: Goat
Clonality: Polyclonal
Clone:
Conjugation: CF®583R
Epitope:
Host: Bovine
Isotype: IgG
Reactivity: Goat
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité