ENZOALX106024G001EA
0
EUR
InStock
ENZOALX106024G001
L-O-Phosphoserine ≥97% (par HPLC)
L-O-Phosphoserine
NMDA receptor antagonist.
L-AP4 agonist. Competitive NMDA antagonist. Has affinity for type IV, but not type I, metabotropic glutamate receptor. Inhibits proliferation. Enhances neuronal differentiation in progenitor cells.
Soluble in water (50 mg/ml).