ENZOBMLSC1060001EA
123.2
EUR
InStock
ENZOBMLSC1060001
Stauprimide ≥98% (par HPLC)
Stauprimide
Potently increases the efficiency of directed differentiation of mouse and human embryonic stem cells in the presence of Activin A (EC₅₀ = 0.3 µM in Sox17+ /Sox7- cells).
Interacts with NME2 and inhibits its nuclear localisation leading to downregulation of c-Myc. Under various conditions, stauprimide treated ESC's were able to be differentiated into endoderm, neuronal, and mesoderm cell lines.
Soluble in DMSO (20 mg/ml).