ENZOALX400048G010EA
63.8
EUR
InStock
ENZOALX400048G010
Busulphan ≥98% agent d'alkylation de l'ADN
Busulphan
Busulfan is antineoplastic bifunctional, DNA alkylating agent that causes DNA damage by cross-linking DNA and proteins.
- Appearance: White to off-white crystalline powder
- Solubility: Soluble in acetone, 100% ethanol or DMSO; insoluble in water