ENZOALX440008M100EA
344.3
EUR
InStock
ENZOALX440008M100
L-Biopterin ≥98%
L-Biopterin
Soluble in 1 N sodium hydroxide or 1 N hydrogen chloride; very slightly soluble in water (0.7 mg/ml) (ultrasonication). To obtain concentrated solutions, ammonium hydroxide should be added.