Lipopolysaccharides (de Salmonella minnesota), TLRgrade™
Fournisseur: ENZO LIFE SCIENCES AG
Danger
ENZOALX581018L002EA 272.8 EUR InStock
ENZOALX581018L002
Lipopolysaccharides (de Salmonella minnesota), TLRgrade™
Lipopolysaccharides
TLR4 activator.
Rough (R)-form LPS isolated and purified from Salmonella minnesota R7 (Rd mutant) by a modification of the PCP extraction method, converted to the uniform sodium salt form.
Strong activator of Toll-like receptor 4 (TLR4). Does not activate TLR2 or other TLRs as determined with splenocytes and macrophages from TLR4 deficient mice. No further re-extraction required. Smooth (S)-form LPS are commonly the preferred choice for whole animal studies, whereas Rough (R)-form LPS are primarily used in cellular in vitro activation studies.
Commander maintenant
Résultats des tests de spécification
Concentration 1 mg/ml
Formulation Liquid. Sterile, ready-to-use solution in pyrogen-free double distilled water
Purity Absence of detectable protein or DNA contaminants with agonistic TLR activity
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité