Mouse Recombinant 4-1BB
Fournisseur: ProSci
PRSI92-374EA 0 EUR InStock
PRSI92-374
Mouse Recombinant 4-1BB
Protéines et Peptides
Tumor necrosis factor receptor superfamily member 9(TNFRSF9) is a member of the tumor necrosis factor (TNF) receptor family. It can be induced by lymphocyte activation (ILA) and is expressed by activated T cells, but to a larger extent on CD8 than on CD4 T cells. In addition, TNFRSF9 expression is found on dendritic cells, follicular dendritic cells, natural killer cells, granulocytes and cells of blood vessel walls at sites of inflammation. As receptor for TNFSF9/4-1BBL, it can activate T cells and the cross-linking of this protein can enhance T cell proliferation, IL-2 secretion survival and cytolytic activity. Further, it can enhance immune activity to eliminate tumors in mice.
  • Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays.
Attention: For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité