Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (iFluor® 710)
Fournisseur: AAT Bioquest
AATB100320K1EA 1390 EUR InStock
AATB100320K1 AATB100320K0
Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (iFluor® 710)
Anticorps
The UCHT1 monoclonal antibody binds to human CD3e, a 20 kDa member of the Ig superfamily commonly found on the surface of tregs, thymocytes, nkt cells and thymocytes (differentiation dependent)s.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Attention: For research use only.
Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: UCHT1
Conjugation: iFluor® 710
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité