Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-CGB Mouse Monoclonal Antibody [clone: HCGb/54]

Fournisseur: ProSci
PRSI33-113EA 0 EUR InStock
PRSI33-113
Anti-CGB Mouse Monoclonal Antibody [clone: HCGb/54]
Anticorps
This antibody reacts with a protein of 22 kDa, identified as beta subunit of HCG. It does not cross react with the alpha subunit. HCG is a glycoprotein which is secreted in large quantities by normal trophoblasts. It is present only in trace amounts in non-pregnant urine and sera but rises sharply during pregnancy. HCG is composed of two non-identical, non-covalently linked polypeptide chains designated as the alpha and beta subunits. The alpha subunit is identical to that of thyroid stimulating hormone (TSH), follicle stimulating hormone (FSH), and luteinizing hormone (LH). HCG-beta antibody detects cells and tumors of trophoblastic origin such as choriocarcinoma. Large cell carcinoma and adenocarcinoma of the lung demonstrate antibody positivity in 90% and 60% of cases respectively; 20% of lung squamous cell carcinomas are positive. HCG expression by non-trophoblastic tumors may indicate aggressive behavior.

ELISA: use BSA-free format (1) IHC (FFPE): 0,5 - 1 µg/ml for 30 min at RT (2) Prediluted format: incubate for 30 min at RT (3) The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the HCG-beta antibody to be titered up or down for optimal performance.<p>1. Clone HCGb/54 (recommended detect) will pair with clones <a href=../tds/hcg-beta-antibody-hcgb211-v2374>HCGb/211</a> (recommended capture) and <a href=../tds/hcg-beta-antibody-hcgb459-v2373>HCGb/459</a> by ELISA.<br>2. Staining of formalin-fixed tissues requires boiling tissue sections in 10 mM citrate buffer, pH 6,0, for 10 - 20 min followed by cooling at RT for 20 minutes.<br>3. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.

Type: Primary
Antigen: CGB
Clonality: Monoclonal
Clone: HCGb/54
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité