Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (iFluor® 790)
Fournisseur: AAT BIOQUEST
100330M0 100330M1
AATB100330M0EA 371 EUR InStock
AATB100330M0 AATB100330M1
Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (iFluor® 790)
Anticorps
The SK7 monoclonal antibody reacts with human CD3e, a 20 kD member of the Ig superfamily often found on the surface of t cells, tregs and nkt cells.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Attention: For research use only.
Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: SK7
Conjugation: iFluor® 790
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité