Human LDL
Fournisseur: STEMCELL Technologies
STMC02698EA 646 EUR InStock
STMC02698
Human LDL
Milieux de culture cellulaire
Low-density lipoproteins pre-screened for hematopoietic cell growth.
This product is a human plasma-derived low-density lipoprotein (LDL)-based culture supplement that has been pre-screened for the culture, expansion, and colony assay of human hematopoietic and non-hematopoietic cells in serum-free culture media. It promotes the proliferation and survival of human hematopoietic and other progenitor cells in culture, resulting in increased cell output in expansion cultures and increased colony numbers and/or size in colony assays.
Commander maintenant
COMMANDE
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité