MethoCult™ H4531 Medium
Fournisseur: STEMCELL Technologies
MethoCult™
STMC04531EA 647 EUR InStock
STMC04531
MethoCult™ H4531 Medium
Milieux de culture cellulaire
Methylcellulose-based medium with agar-LCM (without EPO) for human cells.
MethoCult™ H4531 is methylcellulose-based medium for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of human bone marrow, peripheral blood, mobilised peripheral blood, and cord blood samples. This formulation contains agar leukocyte conditioned medium (agar-LCM) as a source of colony-stimulating factors. MethoCult™ H4531 supports the growth of granulocyte-macrophage progenitor cells (CFU-GM, CFU-G and CFU-M) and EPO-independent erythroid progenitor cells. It is suitable for detection of EPO-independent erythroid progenitor cells.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité