MethoCult™ Starter Kit for H4534 Classic without EPO
Fournisseur: STEMCELL Technologies
MethoCult™
STMC04564EA 1553 EUR InStock
STMC04564
MethoCult™ Starter Kit for H4534 Classic without EPO
Milieux de culture cellulaire
Complete kit for hematopoietic CFU assays.
The starter kit for MethoCult™ H4534 classic without EPO (MethoCult™ GF H4534) is recommended for laboratories in the initial stages of establishing procedures for assessing human hematopoietic progenitor cells using colony-forming unit (CFU) assays. These products support the growth of clonogenic hematopoietic progenitor cells from human bone marrow, peripheral blood, cord blood, leukapheresis products, and purified progenitor-enriched cells. H4534 without EPO is a complete methylcellulose-based medium that supports the growth of granulocyte-macrophage progenitors (CFU-GM, CFU-G and CFU-M). Each kit contains instructional materials in addition to all reagents and materials necessary to perform 24 duplicate assays.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité