NeuroCult™ Differentiation Supplement (Mouse and Rat)
Fournisseur: STEMCELL Technologies
NeuroCult™
STMC05703EA 300 EUR InStock
STMC05703
NeuroCult™ Differentiation Supplement (Mouse and Rat)
Milieux de culture cellulaire
Supplement for differentiation of mouse and rat neural stem and progenitor cells.
NeuroCult™ differentiation supplement (Mouse and Rat) is a standardised, serum-containing supplement for the differentiation of mouse and rat neural stem and progenitor cells into astrocytes, neurons, and oligodendrocytes when used in combination with NeuroCult™ basal medium (Mouse and Rat). This supplement is also a component of NeuroCult™ differentiation kit (Mouse and Rat).
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité