ImmunoCult™ Human Treg Differentiation Supplement
Fournisseur: STEMCELL Technologies
ImmunoCult™
STMC10977EA 279 EUR InStock
STMC10977
ImmunoCult™ Human Treg Differentiation Supplement
Milieux de culture cellulaire
Culture supplement for the differentiation of human naïve CD4+ T cells into tregs.
ImmunoCult™ Human Treg Differentiation Supplement contains a human cytokine and small molecule formulated to promote the robust activation, expansion, and differentiation of peripheral blood-derived, naïve, CD4+ human T cells into regulatory T cells (Tregs). ImmunoCult™ Human Treg Differentiation Supplement is intended for use with ImmunoCult™-XF T Cell Expansion Medium and ImmunoCult™ Human CD3/CD28 T Cell Activator.
This product is recommended for:
Research into the regulation of human naïve T cell differentiation and Treg function
Development of procedures to expand and polarise naïve T cells into Tregs
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité