MethoCult™-GF M3534 Medium
Fournisseur: STEMCELL Technologies
MethoCult™
STMC03534EA 639 EUR InStock
STMC03534
MethoCult™-GF M3534 Medium
Milieux de culture cellulaire
Methylcellulose-based medium with recombinant cytokines (without EPO) for mouse myeloid progenitor cells.
MethoCult™ GF M3534 is optimised for the growth and enumeration of granulocyte-macrophage progenitor cells (CFU-GM, CFU-G, CFU-M) in colony-forming unit (CFU) assays of mouse bone marrow, spleen, peripheral blood, and fetal liver cells. MethoCult™ M3534 does not support the growth of erythroid progenitor cells (BFU-E and CFU-E) as it does not contain erythropoietin (EPO). This formulation is compatible with STEMvision™ software for automated colony counting of mouse bone marrow CFU assays.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité