ClonaCell™-HY Medium
Fournisseur: STEMCELL Technologies
ClonaCell™
STMC03803EA 180 EUR InStock
STMC03803
ClonaCell™-HY Medium
Milieux de culture cellulaire
Hybridoma fusion recovery medium (serum-containing).
ClonaCell™-HY Medium C is a . Serum-containing liquid medium used to promote hybridoma viability and recovery after the fusion process (PEG-based or electrofusion)  and prior to hypoxanthine, aminopterin, thymidine (HAT) selection. This medium has been verified for use in mouse and rat hybridoma development and reportedly is compatible for production of hybridomas using lymphocytes from a variety of host animals including human, mouse, rat, and hamster.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité