STEMdiff™ Cardiomyocyte Maintenance Kit
Fournisseur: STEMCELL Technologies
STEMdiff™
STMC05020EA 205 EUR InStock
STMC05020
STEMdiff™ Cardiomyocyte Maintenance Kit
Milieux de culture cellulaire
Medium for long-term maintenance of human PSC-derived cardiomyocytes.
STEMdiff™ Cardiomyocyte Maintenance Kit can be used for long-term maintenance of human pluripotent stem cell (PSC)-derived cardiomyocytes for one month or longer. The functional capacity of hPSC-derived cardiomyocytes is retained for downstream applications and analysis. This kit is compatible with cardiomyocytes generated using STEMdiff™ Ventricular Cardiomyocyte Differentiation Kit or STEMdiff™ Atrial Cardiomyocyte Differentiation Kit.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité