mTeSR™-1 Medium without Phenol Red
Fournisseur: STEMCELL Technologies
mTeSR™
STMC05876EA 462 EUR InStock
STMC05876
mTeSR™-1 Medium without Phenol Red
Milieux de culture cellulaire
Defined, feeder-free maintenance medium for human ES and iPS cells.
mTeSR™1 is the most widely published feeder-free cell culture medium for human embryonic stem cells (ES cells) and induced pluripotent stem cells (iPS cells), with established protocols for applications ranging from derivation to differentiation. It has been used to successfully maintain hundreds of ES and iPS cell lines in over 40 countries, and has supported top pluripotent stem cell publications and researchers. mTeSR™1 is a highly specialised, serum-free, and complete cell culture medium. With pre-screened raw materials that ensure batch-to-batch consistency and robust feeder-free protocols for ES and iPS cell culture, mTeSR™1 provides more consistent cultures with homogeneous, undifferentiated phenotypes. mTeSR™1 is now available without phenol red, for applications in which the presence of phenol red is undesirable.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité