N2 Supplement-B
Fournisseur: STEMCELL Technologies
STMC07156EA 132 EUR InStock
STMC07156
N2 Supplement-B
Milieux de culture cellulaire
For neural differentiation of mouse and human ES and iPS cells.
N2 Supplement-B, containing iron-poor human apo-transferrin, was developed for the in vitro differentiation of mouse or human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells. N2 Supplement-B can be used in a wide range of protocols to achieve neural induction of human ES/iPS cells and/or downstream differentiation of neural progenitor cells to different neuronal or glial subtypes. N2 Supplement-B is provided as a 100X stock solution.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité