ImmunoCult™ Dendritic Cell Maturation Supplement
Fournisseur: STEMCELL Technologies
ImmunoCult™
STMC10989EA 343 EUR InStock
STMC10989
ImmunoCult™ Dendritic Cell Maturation Supplement
Milieux de culture cellulaire
Supplement for the maturation of human monocyte-derived dendritic cells.
Use ImmunoCult™ Dendritic Cell Maturation Supplement for the maturation of immature dendritic cells (DCs) to mature DCs. For your convenience, the medium is available either individually or as part of the ImmunoCult™ Dendritic Cell Culture Kit. The ImmunoCult™ Dendritic Cell Maturation Supplement is formulated for use in combination with ImmunoCult™-ACF Dendritic Cell Medium and ImmunoCult™-ACF Dendritic Cell Differentiation Supplement to support the maturation of immature DCs.
For more information on protocols for maturation of DCs using ImmunoCult™ Dendritic Cell Maturation Supplement, please explorethe Product Information Sheet (PIS).
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité