StemSpan™ Myeloid Expansion Supplement (100X)
Fournisseur: STEMCELL Technologies
StemSpan™
STMC02693EA 733 EUR InStock
STMC02693
StemSpan™ Myeloid Expansion Supplement (100X)
Milieux de culture cellulaire
Serum-free culture supplement for expansion and differentiation of human granulocytes.
StemSpan™ Myeloid Expansion Supplement (100X) contains a combination of recombinant human cytokines formulated to selectively promote the expansion and differentiation of human myeloid progenitor cells, generating large numbers of CD15+ granulocytes from CD34+ cells isolated from human cord blood (CB) or bone marrow (BM) samples.
StemSpan™ Myeloid Expansion Supplement is intended for use in combination with any of the following StemSpan™ media:
StemSpan™ SFEM
StemSpan™ SFEM II
StemSpan™-XF
StemSpan™-AOF
When added to serum-free medium, StemSpan™ Myeloid Expansion Supplement typically promotes the production of thousands of CD15+ granulocytes per input CD34+ cell in 14-day liquid cultures initiated with CD34+ human CB cells. See data tab for more details.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité