StemSpan™ CC110 Culture Supplement
Fournisseur: STEMCELL Technologies
StemSpan™
STMC02697EA 932 EUR InStock
STMC02697
StemSpan™ CC110 Culture Supplement
Milieux de culture cellulaire
Serum-free culture supplement for expansion of human hematopoietic cells.
StemSpan™ CC110 contains a combination of early-acting recombinant human (rh) cytokines formulated to support the proliferation of human hematopoietic cells. It is supplied as a 100X concentrate. When added to serum-free medium, StemSpan™ CC110 promotes the expansion of CD34+ cells isolated from human cord blood and bone marrow. StemSpan™ CC110 stimulates similar expansion of CD34+ cells compared to StemSpan™ CC100, but with higher purity and is meant for use in short-term cultures to activate stem cell and immature progenitor cycling, without necessarily promoting the proliferation and differentiation of later progenitors.
We recommend using StemSpan™ CC110 in combination with any of the following StemSpan™ media:
StemSpan™ SFEM
StemSpan™ SFEM II
StemSpan™-XF
StemSpan™-AOF
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité