ClonaCell™ TCS/Hygromycin Kit
Fournisseur: STEMCELL Technologies
ClonaCell™
STMC03823EA 640 EUR InStock
STMC03823
ClonaCell™ TCS/Hygromycin Kit
Milieux de culture cellulaire
Kit containing ClonaCell™-TCS Medium and hygromycin B antibiotic.
This kit is meant for selection and cloning applications where the hygromycin B antibiotic is used as the selective agent. ClonaCell™-TCS Medium is a serum-containing, methylcellulose-based semi-solid medium that can be used for selection and cloning of a variety of suspension-adapted and adherent cell lines, such as BHK-1, CHO-K1, CHO-S, CHO-DG44, and HEK-293. This medium contains pre-selected fetal bovine serum (FBS) and bovine serum albumin (BSA). Hygromycin B is suitable for use in cell culture for the selection and maintenance of hygromycin-resistant clones when using ClonaCell™-TCS Medium.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité