MethoCult™ H4035 Optimum without EPO
Fournisseur: STEMCELL Technologies
MethoCult™
STMC04035EA 590 EUR InStock
STMC04035
MethoCult™ H4035 Optimum without EPO
Milieux de culture cellulaire
Methylcellulose-based medium with recombinant cytokines (without EPO) for human cells. 1×100 ml bottle.
MethoCult™ H4035 Optimum Without EPO (MethoCult™ GF H4035) is a methylcellulose-based medium for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of human bone marrow, mobilized peripheral blood, peripheral blood, and cord blood samples. MethoCult™ H4035 Optimum Without is formulated to support optimal growth of granulocyte-macrophage progenitor cells (CFU-GM, CFU-G and CFU-M). The inclusion of G-CSF in Optimum formulations improves discrimination of myeloid colony subtypes. This formulation is compatible with STEMvision™ software for automated colony counting.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité