MyoCult™-SF Expansion Supplement Kit (Human)
Fournisseur: STEMCELL Technologies
MyoCult™
STMC05980EA 603 EUR InStock
STMC05980
MyoCult™-SF Expansion Supplement Kit (Human)
Milieux de culture cellulaire
Serum-free supplement and attachment substrate for the derivation and expansion of human skeletal muscle progenitor cells (myoblasts).
Culture primary human skeletal muscle progenitor cells and human pluripotent stem cell (hPSC)-derived myoblasts under serum-free conditions. The MyoCult™-SF Expansion Supplement Kit (Human) includes components optimised for the derivation and expansion of myoblasts in vitro: MyoCult™-SF Expansion 10X Supplement (Human) and MyoCult™-SF Attachment Substrate. The supplement must be combined with a basal medium (DMEM with 1000 mg/L D-Glucose; sold separately) to prepare MyoCult™-SF Expansion Medium. Use of the MyoCult™-SF Attachment Substrate is optional depending on the cell source used.
For convenience, the kit components may also be purchased individually. If using primary myoblasts, we recommend coating cultureware with MyoCult™-SF Attachment Substrate. For hPSC-derived myoblasts, use Corning® Matrigel® hESC-Qualified Matrix. Myoblasts cultured using MyoCult™-SF Expansion Medium are compatible with MyoCult™ Differentiation Kit (Human) and can be used for further differentiation into multinucleated myotubes. For more information on protocols for derivation and expansion of human skeletal muscle progenitor cells with MyoCult™, please explore the Product Information Sheet.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité