STEMdiff™ Blood Vessel Organoid Maturation Medium
Fournisseur: STEMCELL Technologies
STEMdiff™
STMC100-0658EA 734 EUR InStock
STMC100-0658
STEMdiff™ Blood Vessel Organoid Maturation Medium
Milieux de culture cellulaire
Media for maturation of human pluripotent stem cells derived blood vessels organoids.
Use STEMdiff™ Blood Vessel Organoid Maturation Medium to expand and grow mature blood vessel organoids generated using the STEMdiff™ Blood Vessel Organoid Kit or homemade protocols, with the option to scale up for high-throughput screening in a 96-well format. Compared to homemade organoid culture media formulations, organoids generated with this easy-to-use kit develop extensive and physiologically relevant vascular networks and strong endothelial cell-pericyte interactions. Blood vessel organoids matured using this medium have CD31+/CD34+/CD144+/KDR+ endothelial cells and PDGFR-β+/CD146+/SMA+/NG-2+ pericytes. The simple protocol also eliminates the need for serum screening and labor-intensive protocol optimization.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité