ImmunoCult™ NK Cell Expansion Supplement
Fournisseur: STEMCELL Technologies
ImmunoCult™
STMC100-0715EA 119 EUR InStock
STMC100-0715
ImmunoCult™ NK Cell Expansion Supplement
Milieux de culture cellulaire
Serum-free and animal component-free supplement for expansion of NK cells.
Consistently expand natural killer (NK) cells in serum-free conditions without the use of problematic feeder cells. Use ImmunoCult™ NK Cell Expansion Kit to provide optimised culture conditions for expanding NK cells with high yields. This kit comprises ImmunoCult™ NK Cell Base Medium, ImmunoCult™ NK Cell Expansion Supplement, and ImmunoCult™ NK Cell Expansion Coating Material, providing a complete, easy-to-use culture system. After just 14 days in culture, cells can be harvested and used directly in downstream applications. This kit is compatible with many of our other upstream and downstream products. For example, use EasySep™ cell separation kits to isolate NK cells that can then be immediately expanded using ImmunoCult™ NK Cell Expansion Kit.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité