ImmunoCult™ Mouse B Cell Expansion Kit
Fournisseur: STEMCELL Technologies
ImmunoCult™
STMC100-1003EA 846 EUR InStock
STMC100-1003
ImmunoCult™ Mouse B Cell Expansion Kit
Milieux de culture cellulaire
Serum-free culture kit for in vitro expansion of mouse B cells.
Consistently expand mouse B cells in serum-free conditions without the use of problematic feeder cells. Use ImmunoCult™ Mouse B Cell Expansion Kit to provide optimised culture conditions for expanding B cells from mouse splenocytes with high yields. This kit comprises ImmunoCult™-XF B Cell Base Medium and ImmunoCult™ACF Mouse B Cell Expansion Supplement, providing a robust and easy-to-use culture system. After just 6 - 9 days in culture, cells can be harvested and used directly in downstream applications.
This kit is compatible with our other upstream and downstream products. For example, isolate B cells with EasySep™ Mouse Pan-B Cell Isolation Kit and expand them immediately using ImmunoCult™ Mouse B Cell Expansion Kit.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité