HPV16 (E7) Peptide Pool
Fournisseur: STEMCELL Technologies
STMC100-1394EA 524 EUR InStock
STMC100-1394
HPV16 (E7) Peptide Pool
Protéines et Peptides
Stimulating T cells with HPV16 (E7) Peptide pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. HPV16 (E7) Peptide pool is a lyophilised mixture of 22 peptides from protein E7 of human papillomavirus type 16 (HPV16) consisting of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 98 on protein E7. E7 is an oncoprotein that binds and mediates degradation of the retinoblastoma tumor suppressor protein pRB (Gonzalez et al.). E7 together with E6 demonstrated the ability to immortalize human keratinocytes in vitro, which likely has significance for HPVs oncogenic potential (Hawley-Nelson et al.). Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité