Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Nous tenons à vous informer que notre plateforme d’e-commerce fera l’objet d’une procédure de maintenance planifiée entre 6:00 et 14:00 ET le 26 janvier 2025. Au cours de cette période, notre site ne sera pas disponible. Merci de votre compréhension.

Anti-CD3 Mouse Monoclonal Antibody [clone: OKT3] (APC)

Fournisseur: STEMCELL Technologies
100-0288 100-0289
STMC100-0288EA 131 EUR InStock
STMC100-0288 STMC100-0289
Anti-CD3 Mouse Monoclonal Antibody [clone: OKT3] (APC)
Anticorps
Mouse monoclonal IgG2a, antibody against human CD3, APC-conjugated.

  • T cells

The OKT3 antibody reacts with an extracellular conformational epitope on the ~20 kDa CD3ε subunit of the human T cell receptor (TCR)/CD3 complex, which is expressed on the surface of ~95% of mature T cells and NKT cells, and variably on thymocytes. A majority of T cell neoplasms also express CD3. CD3 is a dimer assembled from combinations of CD3γ, δ, ε, η, and ζ subunits, which are structurally related type 1 transmembrane proteins and members of the immunoglobulin superfamily. CD3 associates non-covalently with the TCR and is involved in transducing antigen recognition signals into the cytoplasm of T cells and in regulating T cell activation and the expression of the TCR on the cell surface. Mutations in CD3 have been associated with disorders of the immune system, including severe combined immunodeficiency (SCID). The OKT3 antibody recognizes an epitope that is expressed when the CD3ε subunit associates with either CD3γ or CD3δ, and binding of the antibody can induce T cell activation. Because of its immunosuppressive properties in vivo, the OKT3 antibody has proven useful in treating allograft rejection. Clone OKT3 is widely used for phenotyping human T cells, and it has been observed to block the binding of anti-CD3 antibody clones SK7 and UCHT1.

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD3
Clonality: Monoclonal
Clone: OKT3
Conjugation: APC
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité