Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-CD11b Rat Monoclonal Antibody [clone: M1/70] (Pacific Blue™)

Fournisseur: STEMCELL Technologies
60001PB 60001PB.1
STMC60001PBEA 321 EUR InStock
STMC60001PB STMC60001PB.1
Anti-CD11b Rat Monoclonal Antibody [clone: M1/70] (Pacific Blue™)
Anticorps
Rat monoclonal IgG2b antibody against human, mouse, rhesus CD11b, Pacific Blue™-conjugated.

  • Granulocytes
  • Subsets
  • Monocytes

The M1/70 antibody reacts with CD11b, an ~170 kDa type 1 transmembrane glycoprotein which associates non-covalently with CD18 to form the heterodimeric Mac-1 receptor. Through its interactions with ligands such as ICAM-1 (CD54), ICAM-2 (CD102), ICAM-4 (CD242), iC3b, and fibrinogen, Mac-1 functions in several processes, including the adherence of neutrophils and monocytes to stimulated endothelium and phagocytosis of complement-coated particles. CD11b is expressed on the surface of granulocytes, monocytes, NK cells, dendritic cells, tissue macrophages, and subsets of T and B cells, and has been used as a marker to distinguish naïve and memory CD8+ T cells. CD11b is a relatively late marker for myeloid differentiation and is undetectable on most myelomonocytic hematopoietic progenitor cells and more primitive cells. The M1/70 antibody reportedly blocks iC3b binding to Mac-1. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse Monocyte Isolation Kit (Catalog #19861).

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD11b
Clonality: Monoclonal
Clone: M1/70
Conjugation: Pacific Blue™
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Baboon, Chimpanzee, Human, Rabbit, Rhesus monkey

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité