Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (APC)

Fournisseur: STEMCELL Technologies
60005AZ 60005AZ.1
STMC60005AZEA 223 EUR InStock
STMC60005AZ STMC60005AZ.1
Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (APC)
Anticorps
Mouse monoclonal IgG1 antibody against human, chimpanzee CD19, APC-conjugated.

  • B cells

The HIB19 antibody reacts with CD19, an ~95 kDa type 1 transmembrane glycoprotein expressed on the surface of B cells throughout all stages of development, from early pre-B cells to plasma cells. Expression is down-regulated but persists in terminally differentiated plasma cells. CD19 is also found on follicular dendritic cells. By associating with CD21 and CD81, CD19 functions as a co-receptor for the B cell receptor and is involved in B cell activation and differentiation. Activation of CD19 is accompanied by phosphorylation of the cytoplasmic domain, which promotes binding to kinases and the induction of intracellular signaling cascades. Mutations in CD19 can result in severe immunodeficiency syndromes. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Human CD19 Positive Selection Kit (Catalog #18054), EasySep™ Human Whole Blood CD19 Positive Selection Kit (Catalog #18084), EasySep™ HLA Whole Blood B Cell Positive Selection Kit (Catalog #18184HLA); partial blocking may be observed, as well as EasySep™ HLA B Cell Enrichment: Complete Processing Kit for Whole Blood (Catalog #19954HLA) and EasySep™ HLA Total Lymphocyte Enrichment: Complete Processing Kit for Whole Blood (Catalog #19961HLA).

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD19
Clonality: Monoclonal
Clone: HIB19
Conjugation: APC
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Chimpanzee, Human

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité