Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-CD41 Mouse Monoclonal Antibody [clone: HIP8] (PE)

Fournisseur: STEMCELL Technologies
60114PE 60114PE.1
STMC60114PEEA 293 EUR InStock
STMC60114PE STMC60114PE.1
Anti-CD41 Mouse Monoclonal Antibody [clone: HIP8] (PE)
Anticorps
Mouse monoclonal IgG1 antibody against human, rhesus, cynomolgus CD41, PE-conjugated.

  • Megakaryocytes
  • Platelets

The HIP8 antibody reacts with the ~125 kDa GPIIbα subunit of human CD41, a heterodimeric integrin expressed primarily on platelets, megakaryocytes, monocytes, and acute megakaryoblastic leukemia cells. The GPIIbα subunit is linked to a smaller ~25 kDa subunit (GPIIbβ) by a single disulfide bond. Three isoforms of CD41 that are generated by alternative splicing have been identified, one of which is expressed only by cancerous cells. CD41 associates non-covalently with CD61 (integrin b3) in a calcium-dependent manner to form the CD41/CD61 (or GPIIb/IIIa) complex, which has been identified as a receptor for several ligands, including fibrinogen, fibronectin, vitronectin, and thrombin. Binding to the CD41/CD61 complex involves an RGD tripeptidyl sequence in the ligand, and both ligand occupancy and ligand-mediated receptor clustering are functionally important in the integrin-mediated response. The HIP8 antibody reportedly blocks platelet aggregation and inhibits the activation of platelets by several compounds.

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD41
Clonality: Monoclonal
Clone: HIP8
Conjugation: PE
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Baboon, Capuchin monkey, Chimpanzee, Cynomolgus monkey, Human, Pig, Rhesus monkey

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité