Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-CD3 Mouse Monoclonal Antibody [clone: UCHT1] (FITC)

Fournisseur: STEMCELL Technologies
60011FI 60011FI.1 100-0447
STMC60011FIEA 151 EUR InStock
STMC60011FI STMC60011FI.1 STMC100-0447
Anti-CD3 Mouse Monoclonal Antibody [clone: UCHT1] (FITC)
Anticorps
Mouse monoclonal IgG1 antibody against human, chimpanzee CD3, FITC-conjugated.

  • T cells

The UCHT1 antibody reacts with the ~20 kDa CD3ε subunit of the human T cell receptor (TCR)/CD3 complex, which is expressed on the surface of ~95% of mature T cells and NKT cells, and variably on thymocytes. A majority of T cell neoplasms also express CD3. The CD3 complex, which is assembled from combinations of CD3γ, δ, ε, η, and ζ subunits, associates non-covalently with the TCR and is involved in transducing antigen recognition signals into the cytoplasm of T cells and in regulating the cell surface expression of the TCR. Activation of T cells by the TCR involves the cytoplasmic tails of the CD3 subunits, which are structurally related type 1 transmembrane proteins and members of the immunoglobulin superfamily. Mutations in the CD3 subunits have been associated with various immunodeficiency disorders including severe combined immunodeficiency (SCID). This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Direct human T cell isolation kit (Catalog #19661), EasySep™ human CD3 positive selection kit II (Catalog #17851; partial blocking may be observed), EasySep™ HLA whole blood T cell enrichment kit (Catalog #19951HLA) and EasySep™ HLA whole blood CD2 positive selection kit (Catalog #18687HLA).

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type: Anti-CD3 Mouse Monoclonal Antibody [clone: UCHT1] (FITC)
Antigen: CD3
Clonality: Monoclonal
Clone: UCHT1
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Chimpanzee, Human

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité