Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-CD267 (TACI) Rat Monoclonal Antibody [clone: 8F10] (APC)

Fournisseur: STEMCELL Technologies
60116AZ 60116AZ.1
STMC60116AZEA 251 EUR InStock
STMC60116AZ STMC60116AZ.1
Anti-CD267 (TACI) Rat Monoclonal Antibody [clone: 8F10] (APC)
Anticorps
Rat monoclonal IgG2a antibody against mouse CD267 (TACI), APC-conjugated.

  • B cells

The 8F10 antibody reacts with mouse CD267 (transmembrane activator and CAML interactor, or TACI), an ~30 kDa type III transmembrane protein of the TNF receptor superfamily that is expressed primarily on the surface of maturing subsets of splenic B cells, including transition type 2 (T2) and marginal zone (MZ) B cells, as well as on plasma cells and plasmablasts. Expression of CD267 is upregulated on B cells exposed to LPS both in vitro and in vivo. CD267 plays important roles in humoral immunity by regulating B cell survival, proliferation, and maturation through its interaction with the ligands APRIL and BAFF. Ligand binding stimulates activation of several transcription factors, including NF-kappa-B, and has been shown to induce either proliferation or apoptosis, suggesting a dual role in the regulation of B cells. CD267 also appears to have an essential function in immunoglobulin isotype switching and B cell differentiation in response to thymus-independent antigens.

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD267 (TACI)
Clonality: Monoclonal
Clone: 8F10
Conjugation: APC
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité