Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-CD117 Rat Monoclonal Antibody [clone: 2B8] (Biotin)

Fournisseur: STEMCELL Technologies
60025BT 60025BT.1
STMC60025BTEA 368 EUR InStock
STMC60025BT STMC60025BT.1
Anti-CD117 Rat Monoclonal Antibody [clone: 2B8] (Biotin)
Anticorps
Rat monoclonal IgG2b antibody against mouse CD117 (c-Kit), biotin-conjugated.

  • Hematopoietic Stem
  • Progenitor cells

The 2B8 antibody reacts with CD117 (c-Kit), an ~145 kDa type 1 transmembrane receptor for c-Kit ligand (stem cell factor/steel factor) that is broadly expressed on hematopoietic stem cells in bone marrow, including pluripotent and erythroid progenitor cells and B and T lymphocyte precursors, as well as on mast cells. CD117 belongs to the tyrosine kinase receptor family and possesses five immunoglobulin-like C2-type domains and a cytoplasmic protein kinase domain. Binding of c-Kit ligand to CD117 induces dimerization and autophosphorylation, which activates several intracellular signaling pathways critical for the proliferation and differentiation of hematopoietic stem cells. Signaling by CD117 is modulated by phosphatases and by rapid endocytosis and degradation of the receptor. The 2B8 antibody does not block binding of c-Kit ligand. Mutations in CD117 are associated with various types of tumors and the piebald trait, an autosomal dominant abnormality of pigmentation. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse Hematopoietic Progenitor Cell Isolation Kit (Catalog #19856).

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD117
Clonality: Monoclonal
Clone: 2B8
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité