Mouse IgA ELISA Antibody Pair Kit
Fournisseur: STEMCELL Technologies
STMC01996EA 543 EUR InStock
STMC01996
Mouse IgA ELISA Antibody Pair Kit
Tests Tests ELISA
For detection and measurement of mouse immunoglobulin A.
The Mouse Immunoglobulin A (IgA) ELISA antibody pair kit is intended for those who want the flexibility of setting up their own ELISA assay. This kit includes capture and detection antibodies and an IgA standard.
It is designed for the quantitative detection and measurement of total mouse IgA in biological fluids such as serum, plasma, and cell culture supernatants. IgA is predominantly found on the surface of epithelial cells, where it acts as a neutralizing antibody against invading pathogens.
IgA primarily exists as a dimer in secretions such as saliva, tears, colostrum, mucus, sweat, and gastric fluid, but also occurs as a monomer in serum, where it comprises 10.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité